accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OHOF

CAS RN 446-31-1

IUPAC Name: 4-amino-2-fluorobenzoic acid
Synonyms: 2-fluoro-4-aminobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: QHERSCUZBKDVOC-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(O)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results