accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FOO

CAS RN 446-32-2

IUPAC Name: 2-amino-4-fluorobenzoate
Synonyms: 2-amino-4-fluorobenzoate
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H5FNO2
InChi Key: LGPVTNAJFDUWLF-UHFFFAOYSA-M
SMILES: NC1=CC(F)=CC=C1C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results