accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FNOO

CAS RN 446-33-3

IUPAC Name: 4-fluoro-2-methyl-1-nitrobenzene
Synonyms: 4-fluoro-2-methyl-1-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: JHFOWEGCZWLHNW-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)