CAS RN 446-34-4
IUPAC Name:
2-fluoro-4-methyl-1-nitrobenzene
Synonyms:
2-fluoro-4-methyl-1-nitrobenzene
Molecular Weight (g/mol):
155.13
Molecular Formula:
C7H6FNO2
InChi Key:
WZMOWQCNPFDWPA-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C(F)=C1)[N+]([O-])=O
1
–
1
of
1
results
1
3-Fluoro-4-nitrotoluene, 99%
CAS: 446-34-4 Molecular Formula: C7H6FNO2 Molecular Weight (g/mol): 155.13 MDL Number: MFCD00007053 InChI Key: WZMOWQCNPFDWPA-UHFFFAOYSA-N Synonym: 3-fluoro-4-nitrotoluene,benzene, 2-fluoro-4-methyl-1-nitro,2-fluoro-4-methylnitrobenzene,2-fluoro-4-methyl-1-nitro-benzene,3-fluoro-4-nitrobenzene,2-fluor-4-methyl-1-nitrobenzol,pubchem1595,3-fluoro4-nitrotoluene,acmc-1alaq,3-fluoro-4-nitro toluene PubChem CID: 67966 IUPAC Name: 2-fluoro-4-methyl-1-nitrobenzene SMILES: CC1=CC=C(C(F)=C1)[N+]([O-])=O