accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHOHO

CAS RN 446-72-0

IUPAC Name: 5,7-dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Synonyms: genistein genisteol genestein differenol A 4,5,7-trihydroxy iso-flavone prunetol genisterin 7-trihydroxyisoflavone 4',5 sophoricol
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: TZBJGXHYKVUXJN-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)