accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ZnOO(R)OH(S)OH(R)OH(R)OHHOOO(R)OH(S)OH(R)OH(R)OHHO

CAS RN 4468-02-4

IUPAC Name: zinc gluconate
Synonyms: zinc gluconate
Molecular Weight (g/mol): 457.69
Molecular Formula: C12H24O14Zn
InChi Key: ZSKINHGFQOKUDM-IYEMJOQQSA-N
SMILES: [Zn].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)