accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSONCS

CAS RN 4478-93-7

IUPAC Name: 1-isothiocyanato-4-methanesulfinylbutane
Synonyms: sulforaphane sulforafan SFN sulforathane DL-sulforaphane
Molecular Weight (g/mol): 177.28
Molecular Formula: C6H11NOS2
InChi Key: SUVMJBTUFCVSAD-UHFFFAOYNA-N
SMILES: CS(=O)CCCCN=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results