accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiClClCl

CAS RN 4484-72-4

IUPAC Name: trichloro(dodecyl)silane
Synonyms: silane, trichlorododecyl-
Molecular Weight (g/mol): 303.77
Molecular Formula: C12H25Cl3Si
InChi Key: BNCXNUWGWUZTCN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results