accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4499-83-6

IUPAC Name: 4,4'-bis({[1,1'-biphenyl]-4-yl})-1,1'-biphenyl
Synonyms: 4,4'-bis({[1,1'-biphenyl]-4-yl})-1,1'-biphenyl
Molecular Weight (g/mol): 458.6
Molecular Formula: C36H26
InChi Key: ZEMDSNVUUOCIED-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results