accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFHN

CAS RN 450-62-4

IUPAC Name: 7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline
Synonyms: 7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline
Molecular Weight (g/mol): 201.19
Molecular Formula: C10H10F3N
InChi Key: RGZZKZNESVFQKR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC2=C(CCCN2)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)