accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 450412-28-9

IUPAC Name: 1-bromo-3-chloro-2-iodobenzene
Synonyms: 1-bromo-3-chloro-2-iodobenzene
Molecular Weight (g/mol): 317.35
Molecular Formula: C6H3BrClI
InChi Key: GIMVCZMZRZGDTL-UHFFFAOYSA-N
SMILES: ClC1=C(I)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results