accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4505-48-0

IUPAC Name: 2-phenyl-1H-indene
Synonyms: 2-phenyl-1H-indene
Molecular Weight (g/mol): 192.26
Molecular Formula: C15H12
InChi Key: BSBXLZYWGGAVHD-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2C=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results