accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOH2NS

CAS RN 4506-71-2

IUPAC Name: ethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Synonyms: ethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular Weight (g/mol): 225.31
Molecular Formula: C11H15NO2S
InChi Key: CDYVTVLXEWMCHU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC2=C1CCCC2
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)