accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 451-13-8

IUPAC Name: 2-(2,5-dihydroxyphenyl)acetic acid
Synonyms: homogentisic acid homogentisinic acid 2,5-dihydroxy-α-toluate 2,5-dihydroxy-a-toluate homogentisate acid alcapton acid, homogentisic 2,5-dihydroxy-a-toluic acid homogentisinate homogentisic acid (substance)
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: IGMNYECMUMZDDF-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(O)=CC=C1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results