accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 451-82-1

OHOF

CAS RN 451-82-1

IUPAC Name: 2-(2-fluorophenyl)acetic acid
Synonyms: O-fluorophenylacetic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: RPTRFSADOICSSK-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1F
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)