accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH2N

CAS RN 4518-10-9

IUPAC Name: methyl 3-aminobenzoate
Synonyms: methyl 3-aminobenzoate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: VZDNXXPBYLGWOS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(N)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)