CAS RN 4518-94-9
IUPAC Name:
dichloro(methyl)propylsilane
Synonyms:
dichloro(methyl)propylsilane
Molecular Weight (g/mol):
157.11
Molecular Formula:
C4H10Cl2Si
InChi Key:
GNVPGBIHGALKRR-UHFFFAOYSA-N
SMILES:
CCC[Si](C)(Cl)Cl