accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3PCH3CH3H3CBr

CAS RN 4519-28-2

IUPAC Name: tetramethylphosphanium bromide
Synonyms: tetramethylphosphanium bromide
Molecular Weight (g/mol): 171.02
Molecular Formula: C4H12BrP
InChi Key: ZTXFOCMYRCGSMU-UHFFFAOYSA-M
SMILES: [Br-].C[P+](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)