accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 452972-09-7

IUPAC Name: (5-bromopyridin-3-yl)boronic acid
Synonyms: 5-bromopyridin-3-ylboronic acid
Molecular Weight (g/mol): 201.81
Molecular Formula: C5H5BBrNO2
InChi Key: ICCGFOKNFZWCTJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=CC(Br)=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results