CAS RN 453-17-8
CAS RN 453-17-8
IUPAC Name:
(2R)-2,3-dihydroxypropanal
Synonyms:
triose D-(+)-glyceraldehyde D-glyceraldehyde
Molecular Weight (g/mol):
90.08
Molecular Formula:
C3H6O3
InChi Key:
MNQZXJOMYWMBOU-VKHMYHEASA-N
SMILES:
OC[C@@H](O)C=O
1
–
1
of
1
results
1
D-Glyceraldehyde (>80%), TRC
CAS: 453-17-8 Molecular Formula: C3 H6 O3 Molecular Weight (g/mol): 90.08 Synonym: Propanal, 2,3-dihydroxy-, (2R)-,Glyceraldehyde, D- (8CI),Propanal, 2,3-dihydroxy-, (R)-,(2R)-2,3-Dihydroxypropanal,(+)-Glyceraldehyde,(2R)-2,3-Dihydroxypropanal,(R)-(+)-Glyceraldehyde,(R)-2,3-Dihydroxypropanal,(R)-Glyceraldehyde,D-(+)-Glyceraldehyde,D-Glyceraldehyde,D-Glycerose,NSC 91534,Triose IUPAC Name: (2R)-2,3-dihydroxypropanal SMILES: OC[C@@H](O)C=O