accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4530-20-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}acetic acid
Synonyms: [(tert-butoxycarbonyl)amino]acetic acid
Molecular Weight (g/mol): 175.18
Molecular Formula: C7H13NO4
InChi Key: VRPJIFMKZZEXLR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(O)=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)