accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4534-11-6

IUPAC Name: hydrogen 3-methyl-4-(propan-2-yl)aniline chloride
Synonyms: hydrogen 4-isopropyl-3-methylaniline chloride
Molecular Weight (g/mol): 185.7
Molecular Formula: C10H16ClN
InChi Key: VPOAQDSAPBSOLW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)C1=CC=C(N)C=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results