accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HSOHO

CAS RN 454-29-5

IUPAC Name: 2-amino-4-sulfanylbutanoic acid
Synonyms: homocysteine DL-homocysteine (free base) HCY Hcy D,L-homocysteine usaf B-12 D,l-homocysteine Homo-CYS
Molecular Weight (g/mol): 135.18
Molecular Formula: C4H9NO2S
InChi Key: FFFHZYDWPBMWHY-UHFFFAOYNA-N
SMILES: NC(CCS)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)