accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFClBr

CAS RN 454-78-4

IUPAC Name: 2-bromo-1-chloro-4-(trifluoromethyl)benzene
Synonyms: 2-bromo-1-chloro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: APSISOSWYXCEQX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C(Br)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)