accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFFF

CAS RN 454-90-0

IUPAC Name: 1-methoxy-3-(trifluoromethyl)benzene
Synonyms: 1-methoxy-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: XHONYVFDZSPELQ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)