accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4546-70-7

IUPAC Name: 5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: adenosine, 2-amino-2'-deoxy-
Molecular Weight (g/mol): 266.26
Molecular Formula: C10H14N6O3
InChi Key: NOLHIMIFXOBLFF-UHFFFAOYNA-N
SMILES: NC1=NC(N)=C2N=CN(C3CC(O)C(CO)O3)C2=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)