accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4546-72-9

IUPAC Name: N-{9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
Synonyms: adenosine, N-benzoyl-2'-deoxy-
Molecular Weight (g/mol): 355.35
Molecular Formula: C17H17N5O4
InChi Key: PIXHJAPVPCVZSV-YNEHKIRRSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results