accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONHNCH3

CAS RN 454712-26-6

IUPAC Name: tert-butyl 3-(methylamino)pyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-(methylamino)pyrrolidine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: OKUCEQDKBKYEJY-UHFFFAOYNA-N
SMILES: CNC1CCN(C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results