accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 455-16-3

IUPAC Name: 4-methylbenzene-1-sulfonyl fluoride
Synonyms: P-fluorosulfonyltoluene
Molecular Weight (g/mol): 174.19
Molecular Formula: C7H7FO2S
InChi Key: IZZYABADQVQHLC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(F)(=O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results