accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OPH

CAS RN 4559-70-0

IUPAC Name: oxodiphenyl-λ⁵-phosphanylium
Synonyms: diphenylphosphine oxide
Molecular Weight (g/mol): 201.18
Molecular Formula: C12H10OP
InChi Key: YFPJFKYCVYXDJK-UHFFFAOYSA-N
SMILES: O=[P+](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)