CAS RN 456-24-6
IUPAC Name:
2-fluoro-5-nitropyridine
Synonyms:
2-fluoro-5-nitropyridine
Molecular Weight (g/mol):
142.09
Molecular Formula:
C5H3FN2O2
InChi Key:
XOZAJNLUAODXSP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(F)N=C1
1
–
1
of
1
results
1
2-Fluoro-5-nitropyridine, 98%
CAS: 456-24-6 Molecular Formula: C5H3FN2O2 Molecular Weight (g/mol): 142.089 MDL Number: MFCD03095059 InChI Key: XOZAJNLUAODXSP-UHFFFAOYSA-N Synonym: 6-fluoro-3-nitropyridine,pyridine, 2-fluoro-5-nitro,2-fluor-5-nitropyridin,2-fluoro-5-nitro-pyridine,abbypharma ap-15-5253,pubchem1202,acmc-1arnr,5-nitro-2-fluoropyridine,2-fluoro-5-nitro pyridine PubChem CID: 95264 IUPAC Name: 2-fluoro-5-nitropyridine SMILES: C1=CC(=NC=C1[N+](=O)[O-])F