CAS RN 4569-86-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methylene Violet 3RAX (~90%), TRC
CAS: 4569-86-2 Molecular Formula: C22H23N4 . Cl Molecular Weight (g/mol): 378.9 Synonym: 3-Amino-7-(diethylamino)-5-phenylphenazin-5-ium Chloride,C.I. 50206,Diethylsafranin,3-Amino-7-diethylamino-5-phenylphenazinium Chloride IUPAC Name: 3-amino-7-(diethylamino)-5-phenylphenazin-5-ium chloride SMILES: CCN(C1=CC2=[N+](C3=CC=CC=C3)C4=CC(N)=CC=C4N=C2C=C1)CC.[Cl-]