accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHOOHOH3CHO(E)(Z)(E)

CAS RN 458-37-7

IUPAC Name: (1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
Synonyms: tumeric merita earth curcumin stain (substance) zlut prirodni 3 yellow root yo-kin curcumin I indian turmeric turmeric (>98% curcurmin) CI natural yellow 3
Molecular Weight (g/mol): 368.39
Molecular Formula: C21H20O6
InChi Key: ZIUSSTSXXLLKKK-KOBPDPAPSA-N
SMILES: COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Quantity 
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)