accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BNBrHOOH

CAS RN 458532-94-0

IUPAC Name: (2-bromopyridin-4-yl)boronic acid
Synonyms: 2-bromopyridin-4-ylboronic acid
Molecular Weight (g/mol): 201.81
Molecular Formula: C5H5BBrNO2
InChi Key: DFIKFQMGZNTFLI-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Br)=NC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results