accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 459-03-0

CH3OF

CAS RN 459-03-0

IUPAC Name: 1-(4-fluorophenyl)propan-2-one
Synonyms: 1-(4-fluorophenyl)propan-2-one
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: ZUEKIIWSVFBTCM-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results