accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 459-60-9

IUPAC Name: 1-fluoro-4-methoxybenzene
Synonyms: P-fluoroanisole
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: VIPWUFMFHBIKQI-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results