accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOF

CAS RN 459-72-3

IUPAC Name: ethyl 2-fluoroacetate
Synonyms: ethyl 2-fluoroacetate
Molecular Weight (g/mol): 106.1
Molecular Formula: C4H7FO2
InChi Key: VCYZVXRKYPKDQB-UHFFFAOYSA-N
SMILES: CCOC(=O)CF
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)