accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHGeH2HClCH3H3C

CAS RN 4593-81-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 261.79
Molecular Formula: C8H22Cl2Ge
InChi Key: MRGRQNCCBMIDLM-UHFFFAOYSA-N
SMILES: Cl.Cl.CCCC[GeH2]CCCC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)