accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4594-45-0

IUPAC Name: 5-(6-chloro-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: 5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 270.67
Molecular Formula: C10H11ClN4O3
InChi Key: PGEULCIODBNODW-UHFFFAOYNA-N
SMILES: OCC1OC(CC1O)N1C=NC2=C(Cl)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results