accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOONN

CAS RN 4595-61-3

IUPAC Name: pyrimidine-5-carboxylic acid
Synonyms: pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: IIVUJUOJERNGQX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CN=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results