accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 46006-36-4

HNNOHO

CAS RN 46006-36-4

IUPAC Name: 1H-1,3-benzodiazole-4-carboxylic acid
Synonyms: 1H-1,3-benzodiazole-4-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: VVQNAFBGAWCMLU-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2N=CNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results