accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NH2NN

CAS RN 461-58-5

IUPAC Name: N''-cyanoguanidine
Synonyms: 2-cyanoguanidine
Molecular Weight (g/mol): 84.08
Molecular Formula: C2H4N4
InChi Key: QGBSISYHAICWAH-UHFFFAOYSA-N
SMILES: NC(N)=NC#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)