accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 461-72-3

IUPAC Name: imidazolidine-2,4-dione
Synonyms: hydantoin
Molecular Weight (g/mol): 100.08
Molecular Formula: C3H4N2O2
InChi Key: WJRBRSLFGCUECM-UHFFFAOYSA-N
SMILES: O=C1CNC(=O)N1
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results