accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 46118-95-0

CH3NSOHON

CAS RN 46118-95-0

IUPAC Name: 2-[(4-methylpyrimidin-2-yl)sulfanyl]acetic acid
Synonyms: [(4-methylpyrimidin-2-yl)sulfanyl]acetic acid
Molecular Weight (g/mol): 184.21
Molecular Formula: C7H8N2O2S
InChi Key: CGMJQRAIQBSTCC-UHFFFAOYSA-N
SMILES: CC1=CC=NC(SCC(O)=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results