accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOSHSH

CAS RN 462-20-4

IUPAC Name: 6,8-disulfanyloctanoic acid
Synonyms: dihydrolipoic acid DHLA dihydro-thioctic acid sodium salt dihydrolipoic acid dihydrolipoic acid, sodium salt dihydro-α-lipoic acid (+-)-isomer dihydrolipoic acid reduced lipoic acid usaf XR-12 6,8-dihydrothioctic acid
Molecular Weight (g/mol): 208.33
Molecular Formula: C8H16O2S2
InChi Key: IZFHEQBZOYJLPK-UHFFFAOYNA-N
SMILES: OC(=O)CCCCC(S)CCS
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results