CAS RN 4620-67-1
IUPAC Name:
1-phenylcyclobutane-1-carbonyl chloride
Synonyms:
1-phenylcyclobutane-1-carbonyl chloride
Molecular Weight (g/mol):
194.66
Molecular Formula:
C11H11ClO
InChi Key:
SVSUIHVKXBTVBI-UHFFFAOYSA-N
SMILES:
ClC(=O)C1(CCC1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Phenylcyclobutanecarbonyl chloride, 97%, Thermo Scientific™
CAS: 4620-67-1 Molecular Formula: C11H11ClO Molecular Weight (g/mol): 194.658 MDL Number: MFCD09903688 InChI Key: SVSUIHVKXBTVBI-UHFFFAOYSA-N Synonym: 1-phenylcyclobutanecarbonyl chloride,1-phenylcyclobutane-carbonyl chloride,cyclobutanecarbonylchloride, 1-phenyl,1-phenylcyclobutancarbonyl chloride,1-chlorocarbonyl-1-phenylcyclobutane PubChem CID: 23294892 IUPAC Name: 1-phenylcyclobutane-1-carbonyl chloride SMILES: C1CC(C1)(C2=CC=CC=C2)C(=O)Cl