CAS RN 4626-22-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydroformononetin, TRC
CAS: 4626-22-6 Molecular Formula: C16 H14 O4 Molecular Weight (g/mol): 270.281 IUPAC Name: 7-hydroxy-3-(4-methoxyphenyl)chroman-4-one SMILES: COc1ccc(cc1)C2COc3cc(O)ccc3C2=O