accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClO

CAS RN 4635-59-0

IUPAC Name: 4-chlorobutanoyl chloride
Synonyms: butanoyl chloride, 4-chloro-
Molecular Weight (g/mol): 140.99
Molecular Formula: C4H6Cl2O
InChi Key: CDIIZULDSLKBKV-UHFFFAOYSA-N
SMILES: ClCCCC(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)