accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3CH3H3C

CAS RN 464-06-2

IUPAC Name: 2,2,3-trimethylbutane
Synonyms: triptane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: ZISSAWUMDACLOM-UHFFFAOYSA-N
SMILES: CC(C)C(C)(C)C
Boiling Point 
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results