accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 464-49-3

H3CH3C(S)(R)CH3O

CAS RN 464-49-3

IUPAC Name: (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: (+-)-camphor caswell no. 155 norcamphor, 1,7,7-trimethyl- camphor, (1r)-isomer synthetic camphor camphor, synthetic camphor usp camphor, (1s)-isomer epa pesticide chemical code 015602 (1s)-camphor
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: DSSYKIVIOFKYAU-XVKPBYJWSA-N
SMILES: CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2
Molecular Weight (g/mol) 
  • (2)
  • (5)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)